Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NELF Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281759AF532
This item is not returnable.
View return policy
Description
NELF Polyclonal antibody specifically detects NELF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NELF | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
nasal embryonic LHRH factorMGC125369, nasal embryonic luteinizing hormone-releasing hormone factor | |
Rabbit polyclonal NELF antibody was raised against a 17 amino acid peptide near the center of human NELF. The immunogen is located within amino acids 230 - 280 of NELF. Amino Acid Squence: RGYAERKRRKRENDSAS | |
0.1 mL | |
Neuroscience | |
26012 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction