Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NELF-E Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105978AF750
Description
NELF-E Polyclonal antibody specifically detects NELF-E in Human samples. It is validated for Western BlotSpecifications
NELF-E | |
Polyclonal | |
Western Blot | |
D6S45, negative elongation factor E, negative elongation factor polypeptide E, NELFE, NELF-ERDmajor histocompatibility complex gene RD, nuclear protein, RD RNA binding protein, RD RNA-binding protein, RDP, RNA-binding protein RD | |
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human negative elongation factor polypeptide E using the numbering given in entry NP_002895.3 (GeneID 7936). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
7936 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction