Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NELL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587839 Supplier Invitrogen™ Supplier No. PA587839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLLLHVDCNR IYERVIDPPD TNLPPGINLW LGQRNQKHGL FKGIIQDGKI IFMPNGYITQ CPNLNHTCPT CSDFLSLVQG IMDLQELLAK MTAKLNYAET RLSQLENCHC EKTCQVSGLL YRDQDSWVDG DHCRNCTCKS GAVECRRMSC PPLNCSPDSL PVHIAGQCCK VCRPKCIYGG KVLAEGQRIL TKSCRECRGG VLVKITEMCP PLNCSEKDHI LPENQCCRVC RGHNFCAEGP KCGENSECKN WNTKATCECK S; Positive Samples: U-87MG, 293T, Mouse pancreas, Rat brain, Rat small intestine; Cellular Location: Cytoplasm, Nucleus envelope.

This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 6.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NELL1
Gene Accession No. Q2VWQ2, Q62919, Q92832
Gene Alias B230343H07Rik; IDH3GL; l7R6; NELL1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1
Gene Symbols NELL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-420 of human NELL1 (NP_0061482).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338352, 4745, 81733
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.