Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NELL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598721 Supplier Invitrogen™ Supplier No. PA598721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NELL1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NELL1
Gene Accession No. Q92832
Gene Alias B230343H07Rik; IDH3GL; l7R6; NELL1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1
Gene Symbols NELL1
Host Species Rabbit
Immunogen Recombinant Human Protein kinase C-binding protein NELL1 protein (160-420AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4745
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.