Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NELL2 Antibody (13E7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174039F
Description
NELL2 Monoclonal specifically detects NELL2 in Human samples. It is validated for ELISA.Specifications
NELL2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NELL2 | |
Recombinant human NELL2 (30-258aa) purified from E. coli | |
100 μL | |
Neuroscience | |
4753 | |
Human, Mouse | |
Purified |
ELISA | |
130000000 | |
ELISA | |
nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human NELL2 mAb, clone 13E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NELL2 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction