Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NELL2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300073AF488
Description
NELL2 Polyclonal specifically detects NELL2 in Human samples. It is validated for ELISA.Specifications
NELL2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 488 | |
nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
Produced in rabbits immunized with purified, recombinant NELL2 (Q99435; Met1-Leu816). | |
0.1 mL | |
Neuroscience | |
4753 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction