Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NELL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547868
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547868 Supplier Invitrogen™ Supplier No. PA547868
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) NELL2 and less than 3% cross-reactivity with rhNELL1 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

NELL2 is a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. Heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis.This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. An alternative splice variant has been described but its full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NELL2
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Nell2
Gene Accession No. Q61220
Gene Alias A330108N19Rik; LOW QUALITY PROTEIN: protein kinase C-binding protein NELL2; mel91; MEL91 protein; Nell2; NEL-like 2; NEL-like 2 (chicken); nel-like 2 homolog; NEL-like protein 2; Nel-related protein 2; neural EGFL like 2; neural epidermal growth factor-like 2; NRP2; Protein kinase C-binding protein NELL2; R75516
Gene Symbols Nell2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NELL2 Leu25-Leu819.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54003
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.