Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NELL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565156
Description
Immunogen sequence: CYCERTCTMKG TTYREFESWI DGCKNCTCLN GTIQCETLIC PNPDCPLKSA LAYVDGKCCK ECKSICQFQG RTYFEGERNT V Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.
NELL2 is a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. Heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis.This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. An alternative splice variant has been described but its full length sequence has not been determined.
Specifications
NELL2 | |
Polyclonal | |
Unconjugated | |
Nell2 | |
A330108N19Rik; LOW QUALITY PROTEIN: protein kinase C-binding protein NELL2; mel91; MEL91 protein; Nell2; NEL-like 2; NEL-like 2 (chicken); nel-like 2 homolog; NEL-like protein 2; Nel-related protein 2; neural EGFL like 2; neural epidermal growth factor-like 2; NRP2; Protein kinase C-binding protein NELL2; R75516 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4753 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99435 | |
Nell2 | |
Recombinant Human NELL2. Recombinant protein control fragment (Product #RP-105905). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction