Learn More
Invitrogen™ NELL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116910
Description
Positive Samples: Mouse liver, Mouse brain, Mouse kidney, Rat testis, Rat brain Immunogen sequence: LGVDPSLQID VLTELELGES TTGVRQVPGL HNGTKAFLFQ DTPRSIKAST ATAEQFFQKL RNKHEFTILV TLKQTHLNSG VILSIHHLDH RYLELESSGH RNEVRLHYRS GSHRPHTEVF PYILADDKWH KLSLAISASH LILHIDCNKI YERVVEKPST DLPLGTTFWL GQRNNAHGYF KGIMQDVQLL VMPQGFIAQC PDLNRTCPTC NDFHGLVQKI MELQDILAKT SAKLSRAEQR MNRLDQCYCE RTCTMKGTTY REFESWIDGC KNCTCLNGT.
NELL2 is a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. Heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis.This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF) -like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. An alternative splice variant has been described but its full length sequence has not been determined.
Specifications
NELL2 | |
Polyclonal | |
Unconjugated | |
Nell2 | |
A330108N19Rik; LOW QUALITY PROTEIN: protein kinase C-binding protein NELL2; mel91; MEL91 protein; Nell2; NEL-like 2; NEL-like 2 (chicken); nel-like 2 homolog; NEL-like protein 2; Nel-related protein 2; neural EGFL like 2; neural epidermal growth factor-like 2; NRP2; Protein kinase C-binding protein NELL2; R75516 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54003, 81734 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.52 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q61220, Q62918 | |
Nell2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-300 of human NELL2 (NP_0061501). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.