Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEMF Monoclonal Antibody (OTI7E10)
GREENER_CHOICE

Catalog No. PIMA527473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527473 Supplier Invitrogen™ Supplier No. MA527473
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEMF Monoclonal Antibody for IHC (P)

NEMF (SDCG1) plays a role in nuclear export and is expressed in brain, heart, liver, lung, spleen, and skeletal muscle. Also expressed at lower levels in stomach and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEMF
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI7E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NEMF
Gene Accession No. O60524
Gene Alias 1500011I12Rik; 4933405E14Rik; Antigen NY-CO-1; Nemf; nuclear export mediator factor; nuclear export mediator factor Nemf; NY-CO-1; SDCCAG1; Serologically defined colon cancer antigen 1; serologically defined colon cancer antigen 1 homolog
Gene Symbols NEMF
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 869-1076 of NEMF produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9147
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.