Learn More
NEMF Mouse anti-Human, Clone: OTI7E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811464

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Nuclear export mediator factor, NEMF, plays a role in nuclear export. It is expressed in brain, heart, liver, lung, spleen, and skeletal muscle tissue, and expressed at lower levels in stomach and testis. It interacts (via its N-terminus) with XPO1; the N-terminal domain contains a nuclear export signal and the C-terminal domain may interact with cargo proteins. This protein is active in normal lung tissue but is inactive in several lung cancer cell lines.Specifications
NEMF | |
Monoclonal | |
Unconjugated | |
NEMF | |
NY-CO-1, SDCCAG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9147 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
OTI7E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60524 | |
NEMF | |
Human recombit protein fragment corresponding to amino acids 869-1076 of human NEMF produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.