Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEMF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552064 Supplier Invitrogen™ Supplier No. PA552064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPRGGQRVSD NIKKETPFLE VITHELQDFA VDDPHDDKEE QDLDQQGNEE NLFDSLTGQP HPEDVLLFAI PICAPYTTMT NYKYKVKLTP GVQKKGKAAK TALNSFMHSK EATAREKDLF RSVKDTDLSR NI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 21%.

NEMF (SDCG1) plays a role in nuclear export and is expressed in brain, heart, liver, lung, spleen, and skeletal muscle. Also expressed at lower levels in stomach and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEMF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEMF
Gene Accession No. O60524
Gene Alias 1500011I12Rik; 4933405E14Rik; Antigen NY-CO-1; Nemf; nuclear export mediator factor; nuclear export mediator factor Nemf; NY-CO-1; SDCCAG1; Serologically defined colon cancer antigen 1; serologically defined colon cancer antigen 1 homolog
Gene Symbols NEMF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEMF. Recombinant protein control fragment (Product #RP-89062).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.