Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEMF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106461 Supplier Invitrogen™ Supplier No. PA5106461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SDCG1.

NEMF (SDCG1) plays a role in nuclear export and is expressed in brain, heart, liver, lung, spleen, and skeletal muscle. Also expressed at lower levels in stomach and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEMF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NEMF
Gene Accession No. O60524, Q8CCP0
Gene Alias 1500011I12Rik; 4933405E14Rik; Antigen NY-CO-1; Nemf; nuclear export mediator factor; nuclear export mediator factor Nemf; NY-CO-1; SDCCAG1; Serologically defined colon cancer antigen 1; serologically defined colon cancer antigen 1 homolog
Gene Symbols NEMF
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEMF(Accession O60524), corresponding to amino acid residues I894-P944.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100322884, 66244, 9147
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.