Learn More
Invitrogen™ NEMF Polyclonal Antibody

Description
The antibody detects endogenous levels of total NEMF protein.
Specifications
Specifications
| Antigen | NEMF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NEMF |
| Gene Accession No. | O60524, Q8CCP0 |
| Gene Alias | 1500011I12Rik; 4933405E14Rik; Antigen NY-CO-1; Nemf; nuclear export mediator factor; nuclear export mediator factor Nemf; NY-CO-1; SDCCAG1; Serologically defined colon cancer antigen 1; serologically defined colon cancer antigen 1 homolog |
| Gene Symbols | NEMF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.