Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NENF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555410 Supplier Invitrogen™ Supplier No. PA555410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLFTEEELAR YGGEEEDQPI YLAVKGVVFD VTSGKEFYGR GAPYNALTGK DSTRGVAKMS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NENF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NENF
Gene Accession No. Q9UMX5
Gene Alias 1110060M21Rik; cell growth-inhibiting protein 47; cell immortalization-related protein 2; CIR2; NENF; nenf {ECO:0000250; Neudesin; neudesin neurotrophic factor; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; Protein GIG47; SCIRP10; SCIRP10-related protein; secreted protein of unknown function; sp2; SP2 protein; Spinal cord injury related protein 10; spinal cord injury-related protein 10; SPUF; SPUF protein; UniProtKB:Q9CQ45}; wu:fa11a04; wu:fb73f07; zgc:112953; zgc:112953 protein; zgc:152703
Gene Symbols NENF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NENF. Recombinant protein control fragment (Product #RP-94160).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.