Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NENF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555875
Description
Immunogen sequence: LDPADLTHDT TGLTAKELEA LDEVFTKVYK AKYPIVGYTA RRILNEDGSP NLDFKPEDQP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 92%.
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.
Specifications
| NENF | |
| Polyclonal | |
| Unconjugated | |
| NENF | |
| 1110060M21Rik; cell growth-inhibiting protein 47; cell immortalization-related protein 2; CIR2; NENF; nenf {ECO:0000250; Neudesin; neudesin neurotrophic factor; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; Protein GIG47; SCIRP10; SCIRP10-related protein; secreted protein of unknown function; sp2; SP2 protein; Spinal cord injury related protein 10; spinal cord injury-related protein 10; SPUF; SPUF protein; UniProtKB:Q9CQ45}; wu:fa11a04; wu:fb73f07; zgc:112953; zgc:112953 protein; zgc:152703 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UMX5 | |
| NENF | |
| Recombinant protein corresponding to Human NENF. Recombinant protein control fragment (Product #RP-94159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction