Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium boride, 99.5%, Thermo Scientific™

Catalog No. AA3949406
Click to view available options
:
5g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12008-23-0
Quantity 5g
Molecular Formula B6Nd
MDL Number MFCD00151359
Synonym neodymium boride
InChI Key BFDCZOFJVLZAJN-UHFFFAOYSA-N
SMILES [Nd].B12B3[B-]11B4B1[B-]234
IUPAC Name pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide neodymium
Molecular Weight (g/mol) 209.10
PubChem CID 92027446
Formula Weight 209.1
Percent Purity 99.5%
Odor Odorless
Chemical Name or Material Neodymium boride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.