Alfa Aesar™ Neodymium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer: Alfa Aesar™ 13964FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA13964FI

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-00-8
    MDL Number MFCD00011130
    UN Number UN3178
    Chemical Name or Material Neodymium foil, 0.1mm (0.004 in.) thick

    Specifications

    Quantity 50 x 50mm
    Packaging Mylar under Argon
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378c-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7440-00-8
    Health Hazard 2 GHS H Statement
    H260-H228
    In contact with water releases flammable gases which may ignite spontaneously.
    Flammable solid.
    UN Number UN3178
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Weight ≈0.43g/25x25mm
    TSCA TSCA
    Physical Form Foil
    MDL Number MFCD00011130
    Odor Odorless
    Chemical Name or Material Neodymium foil, 0.1mm (0.004 in.) thick
    Assay 99.9%, (REO)

    GHS H Statement
    H260-H228
    In contact with water releases flammable gases which may ignite spontaneously.
    Flammable solid.

    GHS P Statement
    P210-P231+P232-P280-P370+P378c-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit