Learn More
Neodymium(III) acetate hydrate, 99.9% (REO)
Description
Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline |
Quantity | 250 g |
Solubility Information | Soluble in water,moderately soluble in strong mineral acids. |
Formula Weight | 321.38 (Anhydrous) |
Percent Purity | 99.9% |
Assay | (REO) |
Chemical Name or Material | Neodymium (III) acetate hydrate |
Safety and Handling
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.