Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous

Catalog No. ac315070050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 10024-93-8 | Cl3Nd | 250.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10024-93-8
Molecular Formula Cl3Nd
Molecular Weight (g/mol) 250.59
MDL Number MFCD00011132

Specifications

Melting Point 784°C
Color White
CAS Min % 99.85
CAS Max % 100.0
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula Cl3Nd
Merck Index 10, 6295
Solubility Information Solubility in water: 96.7g/100mL (13°C)
Formula Weight 250.59
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Neodymium(III) chloride, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-031-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.