Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. p-7048689
Click to view available options
Quantity:
25 g
100 g

CAS: 13709-42-7 | F3Nd | 201.24 g/mol

Neodymium(III) fluoride, is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-42-7
Molecular Formula F3Nd
Molecular Weight (g/mol) 201.24
MDL Number MFCD00011135
InChI Key XRADHEAKQRNYQQ-UHFFFAOYSA-K
IUPAC Name neodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Nd+3]

Specifications

Melting Point 1410°C
Color Purple
Physical Form Crystalline Lump
Quantity 25 g
Linear Formula NdF3
Solubility Information Soluble in aqueous solutions.
Formula Weight 201.24
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.5 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-253-3
RTECSNumber QO9500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.