Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) iodide, ultra dry, 99.9% (metals basis)

Catalog No. p-7052294
Click to view available options
Quantity:
1 g
5 g

CAS: 13813-24-6

Neodymium(III) iodide is used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, catalysis (diene polymerization), laser materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13813-24-6
MDL Number MFCD00016248

Specifications

Color Black
Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 524.95
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Assay (metals basis)
Chemical Name or Material Neodymium(III) iodide, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-467-7
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.