Neodymium(III) oxide, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 11248A1

Quantity 1kg

 View more versions of this product

Catalog No. AA11248A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-97-9
    Physical Form Powder
    Assay Percent Range 99.9% (REO)
    Molecular Formula Nd2O3
    Formula Weight 336.48
    MDL Number MFCD00011134
    Chemical Name or Material Neodymium(III) oxide

    Specifications

    Formula Weight 336.48
    Quantity 1kg
    Color Yellow
    EINECS Number 215-214-1
    Physical Form Powder
    MDL Number MFCD00011134
    CAS 1313-97-9
    Molecular Formula Nd2O3
    Assay Percent Range 99.9% (REO)
    Appearance Light blue powder
    Chemical Name or Material Neodymium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit