Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III)-oxide, 99.9%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1313-97-9
Nd2O3
336.47
MFCD00011134
1313-97-9
Nd2O3
336.47
MFCD00011134
for ultra trace analysis
CAS Min % 99.85
CAS Max % 100.0
Assay Percent Range 99.90%
Linear Formula ND2O3
Merck Index 14, 6450
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acids
Formula Weight 336.47
Physical Form Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Density 7.2400g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: slightly soluble in acids
Specific Gravity 7.24
Total Trace Metal Impurities 0.1% max
Color Blue
Melting Point 2270.0°C
Quantity 25g
Chemical Name or Material Neodymium(III)-oxide, 99.9%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 215-214-1

RTECSNumber : QP0185000

TSCA : TSCA