Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) oxide, REacton™, 99.997% (REO)

Catalog No. p-7049290
Click to view available options
Quantity:
25 g
100 g

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol

Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2272°C
Physical Form Powder
Quantity 25 g
Merck Index 14,6450
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 336.48
Percent Purity 99.997%
Grade Reacton™
Density 7.24 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) oxide
Show More Show Less
EINECSNumber 215-214-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.