Learn More
Neodymium(III) oxide, REacton™, 99% (REO)
Description
ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst

Specifications
Specifications
Melting Point | 2272°C |
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,6450 |
Solubility Information | Insoluble in water; Soluble in acids. |
Formula Weight | 336.48 |
Percent Purity | 99% |
Grade | Reacton™ |
Density | 7.24 g/mL |
Assay | (REO) |
Show More |
Safety and Handling
EINECSNumber | 215-214-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.