Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neogenin Antibody (RM0124-3G55), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122501AF700
Description
Neogenin Monoclonal specifically detects Neogenin in Mouse samples. It is validated for Western Blot.Specifications
| Neogenin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NEO1 | |
| Mouse recombinant protein of Neogenin | |
| 0.1 mL | |
| 4756 | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0124-3G55 | |
| Western Blot | |
| HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Neogenin. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction