Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neogenin Antibody (RM0124-3G55), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122501AF750
Description
Neogenin Monoclonal specifically detects Neogenin in Mouse samples. It is validated for Western Blot.Specifications
Neogenin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
NEO1 | |
Mouse recombinant protein of Neogenin | |
0.1 mL | |
4756 | |
Mouse | |
IgG2 |
Western Blot | |
RM0124-3G55 | |
Western Blot | |
HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066 | |
Rat | |
Protein A or G purified | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Neogenin. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction