Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Neogenin Antibody (RM0124-3G55), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122501MFV500
Description
Neogenin Monoclonal antibody specifically detects Neogenin in Mouse samples. It is validated for Western BlotSpecifications
| Neogenin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 4756 | |
| Store at 4C in the dark. | |
| IgG2 | 
| Western Blot | |
| RM0124-3G55 | |
| Western Blot | |
| HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066 | |
| Mouse recombinant protein of Neogenin | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            