Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neogenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555694 Supplier Invitrogen™ Supplier No. PA555694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGPLPSAPRD VVASLVSTRF IKLTWRTPAS DPHGDNLTYS VFYTKEGIAR ERVENTSHPG EMQVTIQNLM PATVYIFRVM AQNKHGSG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neogenin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Neo1
Gene Accession No. Q92859
Gene Alias 2610028H22Rik; AI327052; D930014N22Rik; HsT17534; IGDCC2; immunoglobulin superfamily DCC subclass member 2; NEO; Neo1; Neogenin; neogenin 1; neogenin homolog 1; NGN; NGNDKFZp547A066; NTN1R2
Gene Symbols Neo1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Neogenin. Recombinant protein control fragment (Product #RP-95255).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.