Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Neomycin Phosphotransferase II Monoclonal Antibody (4B4D1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515275
Description
MA5-15275 targets NPT in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15275 immunogen is purified recombinant fragment of NPT expressed in E. Coli.
Neomycin phosphotransferase II (nptII) gene is used in selection of transformed organisms. It was initially isolated from the transposon Tn5 that was present in the bacterium strain Escherichia coli K12. The gene codes for the aminoglycoside 3'-phosphotransferase (denoted aph(3')-II or NPTII) enzyme. NPTII is probably the most widely used selectable marker for plant transformation. It is also used in gene expression and regulation studies in different organisms in part because N-terminal fusions can be constructed that retain enzymatic activity. In animal cells, G418 and neomycin are used as selectable agents.
Specifications
Neomycin Phosphotransferase II | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
Purified recombinant fragment of NPT expressed in E. Coli. | |
RUO | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
4B4D1 | |
Unconjugated | |
Mouse | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction