Learn More
Thermo Scientific Chemicals Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Type | Neomycin Sulfate |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Chemical Name or Material | Neomycin sulfate, 50mg/mL in distilled water, sterile-filtered |
Safety and Handling
| Hazard Category | H317-H334-H361d |
| Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c |
| EINECSNumber | 215-773-1 |
| TSCA | Yes |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.