Learn More
Invitrogen™ Neon™ NxT Electroporation System 100-μL Kit with 1-Channel Tubes

Description
The Neon NxT Electroporation System 100-μL Kit with 1-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Use this kit for transfection volumes of 100 μL, containing 5 x 105–2 x 106 adherent cells or 1 x 106–5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be washed and plated into a multi-well culture dish.

Specifications
Specifications
Content And Storage | • 10 mL Resuspension R Buffer; store at 4°C • 10 mL Resuspension T Buffer; store at 4°C • 10 mL Resuspension Genome Editing Buffer; store at 4°C • 50 mL Electrolytic E100 Buffer; store at 4°C • 25 reaction tips • 8 electroporation 1-channel tubes |
Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
Format | Electroporation Pipette Tip |
Transfection Technique | Electroporation |
For Use With (Equipment) | Neon™ NxT Electroporation System |
For Use With (Application) | Transfection |
No. of Reactions | 25 x 2 Reactions |
Product Type | Pipette Tip |
Quantity | 25 x 2 reactions |
Volume (Metric) Sample | 100 μL |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.