Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neon™ NxT Electroporation System 100 μL Kit with 8-Channel Tubes

Catalog No. p-200087436
Encompass_Preferred
Click to view available options
No. of Reactions:
384 x 2 Reactions
96 x 2 Reactions

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.

The Neon NxT Electroporation System 100-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System (Cat. No. MPK5000) with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 100 μL, containing 5 x 105 to 2 x 106 adherent cells or 1 x 106 to 5 x 106 suspension cells. Cells that have been transfected using the included 100-μL Neon NxT tips are then ready to be plated into a multi-well culture dish.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Box 1 includes:
• Resuspension R Buffer, 4 x 10 mL
• Resuspension T Buffer, 4 x 10 mL
• Resuspension Genome Editing Buffer, 4 x 1 mL
• Electrolytic E100 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 96 tips
• Electroporation 8-channel tubes, 8 tubes

Store at room temperature.
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
Transfection Technique Electroporation
For Use With (Equipment) Neon™ NxT Electroporation System
For Use With (Application) Transfection
No. of Reactions 96 x 2 Reactions
Quantity 96 x 2 Reactions
Volume (Metric) Sample 100 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.