Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neon™ Transfection System Pipette

Catalog No. MPP100
Encompass_Preferred
Click to view available options
Quantity:
1 each

Neon™ Transfection System Pipette

The Neon™ Transfection System Pipette is designed specifically for use with the Neon™ Transfection System device (MPK5000). Each device (MPK5000) already comes with a pipette. The pipette is also sold separately for those labs needing and additional pipette or replacement for their existing pipette.

 

For more information on the product, please click here

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The Neon™ Transfection System Pipette comes with 1 x pipette. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions.
For Use With Transfection Grade Nucleic Acid Purification Kits, siRNA Vectors, DNA Vectors
Product Line Neon
Type Electroporation Pipette Tip
For Use With (Equipment) Neon™ Transfection System
Quantity 1 each
Serum Compatible Yes
Shipping Condition Room Temperature
Starting Material Cell No. 2x10^4 - 6 x10^8 cells; Varies by cell type
System Type Neon™ Transfection System
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.