Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyl alcohol, 99%

Catalog No. AC128250500
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-84-3 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-84-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004682
InChI Key KPSSIOMAKSHJJG-UHFFFAOYSA-N
Synonym neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
PubChem CID 6404
IUPAC Name 2,2-dimethylpropan-1-ol
SMILES CC(C)(C)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 52°C to 53°C
Color Colorless
Density 0.8100g/mL
Boiling Point 113°C to 114°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2OH
Quantity 50 g
Beilstein 01, 406
Specific Gravity 0.81
Merck Index 15, 6542
Solubility Information Solubility in water: 3.5% (25 c)
Formula Weight 88.15
Percent Purity 99%
Physical Form Waxy Crystalline Solid
Chemical Name or Material Neopentyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-907-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.