Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Neoprene Hose for Bunsen Burner PROMO
SCI_ED

High quality neoprene tubing to connect Bunsen burners to fuel source.

$18.00

Specifications

Product Type Gas Hose
For Use With (Equipment) Bunsen burner
Gas Type Natural Gas/Propane
Material Neoprene
Color Blue
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
S24487FB25
SCI_ED
View Documents
Eisco™
CH0100BS25
3 ft. x 0.31 to 0.39 in.
Each for $18.00
Only null left
Add to Cart
 
S12817FB25
SCI_ED
View Documents
Eisco™
CH0100AS25
2 ft. x 0.31 to 0.39 in.
Each for $18.00
Only null left
Add to Cart
 
Description

Description

  • Connect your Bunsen burners to a fuel source such as natural gas or liquid propane
  • Both ends of the hose are designed to fit gas inlet diameters of roughly 8 to 10 mm
Specifications

Specifications

Gas Hose
Natural Gas/Propane
Blue
Connect burners to fuel source
Bunsen burner
Neoprene
8 to 10 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.