Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Neoprene Stoppers
SDPSCI_ED

Catalog No. S23951 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Stopper Number:
0
00
000
1
2
3
4
5
6
6.5
7
8
9
10
10.5
11
11.5
12
13
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
S23951 0
S23949 000
S23950 00
S23952 1
S82858 2
S82859 3
S23955 4
S23956 5
S23958 6
S23959 6.5
S82860 7
S82861 8
S23962 9
S23963 10
S82862 10.5
S82863 11
S82864 11.5
S82865 12
S82866 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Prop 65 Warning
Catalog No. S23951 Supplier United Scientific™ Supplier No. NST0S
Add to Cart
Add to Cart
Prop 65 Warning

Used in areas where rubber would be attacked by reagents or solvents. Neoprene Stoppers have good resiliency and shelf-life.

  • Withstand petroleum products and oils as well as most inorganic acids and bases.
  • Green for easy identification.
  • Sold by the pound.

Warranty and Services

1 Year

Specifications

Diameter (Metric) Outer 17 mm
Diameter (Metric) Bottom 13 mm
Shape Tapered
Autoclavable Autoclavable
Diameter (Metric) Top 17 mm
Product Type Tapered Stopper
Stopper Number 0
Color Green
Material Neoprene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.