Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Neoprene Stoppers
SDPSCI_ED

Used in areas where rubber would be attacked by reagents or solvents. Neoprene Stoppers have good resiliency and shelf-life.

Supplier:  United Scientific™ NST4S

 View more versions of this product

Catalog No. S23955


Only null left
Add to Cart

Alert:

Prop 65 Warning


Description

Description

  • Withstand petroleum products and oils as well as most inorganic acids and bases.
  • Green for easy identification.
  • Sold by the pound.

Warranty and Services

1 Year

Specifications

Specifications

26 mm
20 mm
Neoprene
4
Autoclavable
26 mm
Tapered Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.