Learn More
United Scientific™ Neoprene Stoppers
Used in areas where rubber would be attacked by reagents or solvents. Neoprene Stoppers have good resiliency and shelf-life.
$21.25 - $23.75
Specifications
Autoclavable | Autoclavable |
---|---|
Color | Green |
Material | Neoprene |
Product Type | Tapered Stopper |
Shape | Tapered |
Alert:
Prop 65 Warning
Catalog Number | Mfr. No. | Stopper Number | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Stopper Number | Price | Quantity & Availability | |||||
S23949
![]() ![]() |
United Scientific™
NST000S |
000 |
Pack for $23.75
|
|
|||||
S23950
![]() ![]() |
United Scientific™
NST00S |
00 |
Pack for $23.75
|
|
|||||
S23951
![]() ![]() |
United Scientific™
NST0S |
0 |
Pack for $23.75
|
|
|||||
S23952
![]() ![]() |
United Scientific™
NST1S |
1 |
Pack for $23.75
|
|
|||||
S82858
![]() ![]() |
United Scientific™
NST2S |
2 |
Pack of 48 for $21.25
|
|
|||||
S82859
![]() ![]() |
United Scientific™
NST3S |
3 |
Pack of 35 for $21.25
|
|
|||||
S23955
![]() ![]() |
United Scientific™
NST4S |
4 |
Pack for $23.75
|
|
|||||
S23956
![]() ![]() |
United Scientific™
NST5S |
5 |
Pack for $23.75
|
|
|||||
S82860
![]() ![]() |
United Scientific™
NST7S |
7 |
Pack of 14 for $21.25
|
|
|||||
S82861
![]() ![]() |
United Scientific™
NST8S |
8 |
Dozen of 12 for $21.25
|
|
|||||
S23963
![]() ![]() |
United Scientific™
NST10S |
10 |
Pack for $23.75
|
|
|||||
S82862
![]() ![]() |
United Scientific™
NST10AS |
10.5 |
Pack of 7 for $21.25
|
|
|||||
S82863
![]() ![]() |
United Scientific™
NST11S |
11 |
Pack of 6 for $21.25
|
|
|||||
S82864
![]() ![]() |
United Scientific™
NST11AS |
11.5 |
Pack of 6 for $21.25
|
|
|||||
S82865
![]() ![]() |
United Scientific™
NST12S |
12 |
Pack of 5 for $21.25
|
|
|||||
S82866
![]() ![]() |
United Scientific™
NST13S |
13 |
Pack of 4 for $21.75
|
|
|||||
Description
- Withstand petroleum products and oils as well as most inorganic acids and bases.
- Green for easy identification.
- Sold by the pound.
Warranty and Services
1 Year
Specifications
Autoclavable | |
Neoprene | |
Tapered |
Green | |
Tapered Stopper | |
Non-sterile |