Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Neoprene Stoppers



Used in areas where rubber would be attacked by reagents or solvents. Neoprene Stoppers have good resiliency and shelf-life.
Supplier: United Scientific™ NST11S
Alert:
Prop 65 Warning
Description
- Withstand petroleum products and oils as well as most inorganic acids and bases.
- Green for easy identification.
- Sold by the pound.
Warranty and Services
1 Year
Specifications
56 mm | |
Neoprene | |
Autoclavable | |
Tapered |
Tapered Stopper | |
0.5 kg (1 lb.) | |
Green | |
11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction