Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nephrin Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318058FR
This item is not returnable.
View return policy
Description
Nephrin Polyclonal antibody specifically detects Nephrin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Nephrin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor | |
Partial recombinant protein made to the internal portion of human Nephrin (amino acids 23-1029) [UniProt O60500]. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4868 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction