Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nephrin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$201.00 - $425.50
Specifications
| Antigen | Nephrin |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
Nephrin Polyclonal antibody specifically detects Nephrin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Nephrin | |
| Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor | |
| Partial recombinant protein made to the internal portion of human Nephrin (amino acids 23-1029) [UniProt O60500]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS | |
| 4868 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title