Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nephrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591907 Supplier Invitrogen™ Supplier No. PA591907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse kidney, mouse brain, rat brain; Cellular Location: Cell membrane, Single-pass type I membrane protein Immunogen sequence: LGDSGLADKG TQLPITTPGL HQPSGEPEDQ LPTEPPSGPS GLPLLPVLFA LGGLLLLSNA SCVGGVLWQR RLRRLAEGIS EKTEAGSEED RVRNEYEESQ WTGERDTQSS TVSTTEAEPY YRSLRDFSPQ LPPTQEEVSY SRGFTGEDED MAFPGHLYDE VERTYPPSGA WGPLYDEVQM GPWDLHWPED TYQDPRGIYD QVAGDLDTLE PDSLPFELRG HLV.

Primary steroid resistant nephrotic syndrome (SRNS) is characterized by childhood onset of proteinuria and progression to end stage renal disease. Kidney podocytes and their slit diaphragms form the final barrier to urinary protein loss. Congenital nephrotic syndrome (CNS) is caused by mutations in NPHS1 (nephrin) or NPHS2. Nephrin, a recently identified protein is a member of a group of podocyte proteins that constitute major component of the slit diaphragm especially in the foot process. Nephrin, a cell adhesion molecule, may play a crucial role in maintaining the glomerular filtration barrier. Recent studies have suggested that mutations in the gene for Nephrin reportedly lead to congenital nephrosis. Three novel podocyte proteins, Podocin, Nephrin and alpha Actinin 4 have been identified in congenital and experimental models of proteinuria. The role of Nephrin in anti apoptotic activity in podocyte slit diaphragm is believed to be associated with vascular endothelial derived growth factors VEGF signaling. Nephrin seems to play a role in the development or function of the kidney glomerular filtration barrier and it may anchor the podocyte slit diaphragm to the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nephrin
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Nphs1
Gene Accession No. Q9QZS7, Q9R044
Gene Alias CNF; Nephrin; nephrin 1; NephrinB; nephrosis 1 homolog, nephrin; nephrosis 1, congenital, Finnish type (nephrin); nephrosis 1, nephrin; NPHN; NPHS1; NPHS1 nephrin; NPHS1, nephrin; renal glomerulus-specific cell adhesion receptor
Gene Symbols Nphs1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54631, 64563
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.