Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neprilysin 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981104
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1104 Supplier Bioss Supplier No. BS11102R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMEL1 is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. MMEL1 is a type II transmembrane protein and is thought to be expressed as a secreted protein. The protein encoded by this gene is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. This protein is a type II transmembrane protein and is thought to be expressed as a secreted protein. This gene is expressed mainly in testis with weak expression in the brain, kidney, and heart.

Specifications

Antigen Neprilysin 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MMEL1
Gene Accession No. Q495T6
Gene Alias BOS_15998; EC 3.4.24; EC 3.4.24.11; mel transforming oncogene-like 1; MELL1; membrane metalloendopeptidase like 1; Membrane metallo-endopeptidase-like 1; Membrane metallo-endopeptidase-like 1, soluble form; Membrane metallo-endopeptidase-like 2; MGC119454; MGC119455; MGC119456; MMEL1; MMEL2; NEP2; NEP2(m); NEP2(s); NEPII; NEPIIMGC119456; NEPLP; NEPLP alpha; NEPLP beta; NEPLP gamma; neprilysin 2; Neprilysin II; Neprilysin-2; Neprilysin-2 secreted; Neprilysin-like 1; Neprilysin-like peptidase; Nl1; NL-1; NL2; NL2NL1; Sep; Soluble secreted endopeptidase; zinc metallopeptidase
Gene Symbols MMEL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Neprilysin 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79258
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.