Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neprilysin/CD10 Antibody (CB-CALLA), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247961AF532
Description
Neprilysin/CD10 Monoclonal specifically detects Neprilysin/CD10 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
Neprilysin/CD10 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MME | |
Human Neprilysin/CD10 (Uniprot: P08473) | |
0.1 mL | |
Primary | |
Recognizes a 100kDa glycoprotein, identified as CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity, which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt′s, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
CB-CALLA | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence | |
Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase | |
Mouse | |
Protein A or G purified | |
B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
4311 | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction