Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-(2,4-Dinitrophenyl)-L-lysine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. d1023100mg
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 14401-10-6
Molecular Formula C12H16ClN4O6
Molecular Weight (g/mol) 347.73
MDL Number MFCD00036383
InChI Key FJDUWQCVTQNLPA-UHFFFAOYNA-N
Synonym Nepsilon-Dnp-L-lysine Hydrochloride, H-Lys(Dnp)-OH.HCl
PubChem CID 22872935
IUPAC Name 2-amino-6-[(2,4-dinitrophenyl)amino]hexanoic acid hydrochloridyl
SMILES [Cl].NC(CCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 348.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-(2,4-Dinitrophenyl)-L-lysine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.