Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-L-lysinamide hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
58117-53-6
C14H22ClN3O3
315.798
MFCD00035520
QSRGTPRDCDSKAY-YDALLXLXSA-N
h-lys z-nh2.hcl,h-lys z-nh2 hcl,benzyl s-5,6-diamino-6-oxohexyl carbamate monohydrochloride,nepsilon-z-l-lysine amide hydrochloride,h-lys z-nh2cl,h-lys z-nh2 hydrochloride,s-2-amino-6-benzyloxycarbonylamino hexanamide hydrochloride,benzyl n-5s-5,6-diamino-6-oxohexyl carbamate hydrochloride,benzyl n-5s-5,6-diamino-6-oxohexyl carbamate;hydrochloride,benzyl n-5s-5-amino-5-carbamoylpentyl carbamate hydrochloride
6453579
benzyl N-[(5S)-5,6-diamino-6-oxohexyl]carbamate;hydrochloride
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)N)N.Cl
58117-53-6
C14H22ClN3O3
315.798
MFCD00035520
QSRGTPRDCDSKAY-YDALLXLXSA-N
h-lys z-nh2.hcl,h-lys z-nh2 hcl,benzyl s-5,6-diamino-6-oxohexyl carbamate monohydrochloride,nepsilon-z-l-lysine amide hydrochloride,h-lys z-nh2cl,h-lys z-nh2 hydrochloride,s-2-amino-6-benzyloxycarbonylamino hexanamide hydrochloride,benzyl n-5s-5,6-diamino-6-oxohexyl carbamate hydrochloride,benzyl n-5s-5,6-diamino-6-oxohexyl carbamate;hydrochloride,benzyl n-5s-5-amino-5-carbamoylpentyl carbamate hydrochloride
6453579
benzyl N-[(5S)-5,6-diamino-6-oxohexyl]carbamate;hydrochloride
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)N)N.Cl
Beilstein 6088865
Formula Weight 315.8
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysinamide hydrochloride

storageNote1 : 2° to 8°C