Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%

Catalog No. AAH5918206
Click to view available options
Quantity:
5 g

CAS: 71989-26-9 | C26H32N2O6 | 468.55 g/mol

It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD000371
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
Synonym fmoc-lys boc-oh, fmoc-lys boc, na-fmoc-ne-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n-fmoc-n'-boc-l-lysine, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n, a-fmoc-n, a-boc-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 2724628
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 130°C to 135°C
Quantity 5 g
Beilstein 4217767
Solubility Information Slightly soluble in water.
Optical Rotation −12° (c=1 in DMF)
Formula Weight 468.54
Percent Purity 98%
Chemical Name or Material Nepsilon-Boc-Nalpha-Fmoc-L-lysine
EINECSNumber 276-256-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.