Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Neratinib
GREENER_CHOICESDP

Catalog No. 737150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-715-0 50 mg
73-711-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-715-0 Supplier Tocris Bioscience™ Supplier No. 7371/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent HER2 receptor tyrosine kinase and EGFR kinase inhibitor

Neratinib is a potent, irreversible inhibitor of HER-2/ErbB2 receptor tyrosine kinase and EGFR kinase (IC50 = 59 nM and 92 nM, respectively). Neratinib reduces receptor autophosphorylation and proliferation of EGFR-dependent cancer cells, blocks cell-cycle progression and inhibits HER-2 signaling pathways. Neratinib suppresses proliferation of gefitinib -resistant non-small cell lung cancer cells expressing mutant EGFR. Also inhibits MST1 (IC50 = 37.7 nM). Orally bioavailable.

Specifications

Chemical Name or Material (2 E)-N-[4-[[3-Chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide
CAS 698387-09-6
Quantity 50 mg
Target EGF Receptor Inhibitors
Molecular Formula C30H29ClN6O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.